Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dimethylacetanilide, 98%

Chemical Identifiers

Chemical Name or Material 2',4'-Dimethylacetanilide
CAS 2050-43-3
Formula Weight 163.22
MDL Number MFCD00026125
Synonym N-(2,4-Dimethylphenyl)acetamide; 2',4'-Acetoxylidide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5560506

  SDS
alfa aesar™
H5560506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5560514

  SDS
alfa aesar™
H5560514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 129° to 132°C
Formula Weight 163.22
TSCA TSCA
Physical Form Powder
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AN7420000
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 2050-43-3
Chemical Name or Material 2',4'-Dimethylacetanilide
Health Hazard 1 Warning
Quantity 5g
Assay 98%
Synonym N-(2,4-Dimethylphenyl)acetamide; 2',4'-Acetoxylidide
Color White
MDL Number MFCD00026125
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.