Learn More
2',4'-Dimethylacetanilide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5560514
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2',4'-Dimethylacetanilide | |
129°C to 132°C | |
C10H13NO | |
25 g | |
PJEIIBXJUDOMAN-UHFFFAOYSA-N | |
N-(2,4-dimethylphenyl)acetamide | |
16303 | |
98% |
2050-43-3 | |
White | |
MFCD00026125 | |
n-2,4-dimethylphenyl acetamide, 2',4'-dimethylacetanilide, 2,4-dimethylacetanilide, 4-acetamido-m-xylene, 2',4'-acetoxylidine, 2',4'-acetoxylidide, acetamide, n-2,4-dimethylphenyl, acetanilide, 2',4'-dimethyl, n-acetyl-2,4-xylidine, unii-3u0w52hz1h | |
CC1=CC(=C(C=C1)NC(=O)C)C | |
163.22 | |
163.22 | |
Powder |
Chemical Identifiers
2050-43-3 | |
163.22 | |
PJEIIBXJUDOMAN-UHFFFAOYSA-N | |
16303 | |
CC1=CC(=C(C=C1)NC(=O)C)C |
C10H13NO | |
MFCD00026125 | |
n-2,4-dimethylphenyl acetamide, 2',4'-dimethylacetanilide, 2,4-dimethylacetanilide, 4-acetamido-m-xylene, 2',4'-acetoxylidine, 2',4'-acetoxylidide, acetamide, n-2,4-dimethylphenyl, acetanilide, 2',4'-dimethyl, n-acetyl-2,4-xylidine, unii-3u0w52hz1h | |
N-(2,4-dimethylphenyl)acetamide |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 218-088-6
RTECSNumber : AN7420000
TSCA : Yes
Recommended Storage : Ambient temperatures