Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethyl-6-nitroaniline, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dimethyl-6-nitroaniline
CAS 1635-84-3
Molecular Formula C8H10N2O2
Formula Weight 166.18
MDL Number MFCD00007739
UN Number UN2811
Synonym 6-Nitro-2,4-xylidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2592403

 
alfa aesar™
H2592403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2592406

 
alfa aesar™
H2592406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material 2,4-Dimethyl-6-nitroaniline
Assay 97%
Molecular Formula C8H10N2O2
Melting Point 65° to 68°C
MDL Number MFCD00007739
CAS 1635-84-3
UN Number UN2811
Quantity 1g
Synonym 6-Nitro-2,4-xylidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.