Please login to your online account to display your discounted pricing

2,4-Dimethyl-6-hydroxypyrimidine +99%, ACROS Organics™

$34.21 - $34.21

Chemical Identifiers

Chemical Name or Material 2, 4-Dimethyl-6-hydroxypyrimidine
Name Note 99+%
CAS 6622-92-0
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H8N2O
Formula Weight 124.14
MDL Number MFCD00006105
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC116350050

  SDS
thermo fisher scientific
116350050
Glass bottle 5g Each for $34.21

AC116350250

  SDS
thermo fisher scientific
116350250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.14
Assay ≥99.0%
Physical Form Crystalline Needles
Quantity 5g
Color White
EINECS Number 229-577-9
TSCA TSCA
Melting Point 198° to 200°C
CAS Min % 99.0
CAS Max % 100.0
Synonym 2, 4-Dimethyl-6-pyrimidinol
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
Chemical Name or Material 2, 4-Dimethyl-6-hydroxypyrimidine
Molecular Formula C6H8N2O
MDL Number MFCD00006105
CAS 6622-92-0