missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethyl-3-pentanone, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 159705000

 View more versions of this product

Catalog No. AC159705000



Specifications

Specifications

2, 4-Dimethyl-3-pentanone
15°C
Glass bottle
0.8
124°C
-80°C
95%
97.5
97.5% min. (GC)
(CH3)2CHCOCH(CH3)2
01, 703
Solubility in water: insoluble
CC(C)C(=O)C(C)C
114.19
114.19
98%
0.8000g/mL
Authentic
1.3990 to 1.402
0.63 mPa.s (20°C)
Colorless to Yellow
500mL
565-80-0
100.0
C7H14O
MFCD00008918
2,4-dimethyl-3-pentanone
HXVNBWAKAOHACI-UHFFFAOYSA-N
2,4-dimethylpentan-3-one
11271
Liquid

Chemical Identifiers

565-80-0
114.19
HXVNBWAKAOHACI-UHFFFAOYSA-N
11271
CC(C)C(=O)C(C)C
C7H14O
MFCD00008918
2,4-dimethyl-3-pentanone
2,4-dimethylpentan-3-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-294-7

RTECSNumber : SA8575500

TSCA : TSCA

SDS Documents