missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethyl-3-pentanone 95%, ACROS Organics™

Manufacturer:  Acros Organics 159702500

 View more versions of this product

Catalog No. AC159702500



Specifications

Specifications

2, 4-Dimethyl-3-pentanone
Authentic
1.3990 to 1.4020 (20°C, 589nm)
250mL
565-80-0
100.0
C7H14O
MFCD00008918
2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
CC(C)C(=O)C(C)C
114.188
114.19
≥97.5%
0.8000g/mL
Glass bottle
Colorless to Yellow
95%
97.5
98%
(CH3)2CHCOCH(CH3)2
01, 703
HXVNBWAKAOHACI-UHFFFAOYSA-N
2,4-dimethylpentan-3-one
11271
Liquid

Chemical Identifiers

565-80-0
114.188
HXVNBWAKAOHACI-UHFFFAOYSA-N
11271
CC(C)C(=O)C(C)C
C7H14O
MFCD00008918
2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
2,4-dimethylpentan-3-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-294-7

RTECSNumber : SA8575500

TSCA : TSCA

SDS Documents