Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethyl-3-pentanol, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dimethyl-3-pentanol
CAS 600-36-2
Molecular Formula C7H16O
Formula Weight 116.2
MDL Number MFCD00008921
UN Number UN1987
Beilstein 1731593
Synonym Diisopropyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2082114

  SDS
alfa aesar™
B2082114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.2
Chemical Name or Material 2,4-Dimethyl-3-pentanol
TSCA TSCA
Assay 97%
Molecular Formula C7H16O
MDL Number MFCD00008921
CAS 600-36-2
UN Number UN1987
Beilstein 1731593
Synonym Diisopropyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number SA6660000
Flash Point 37°C (99°F)
Odor Characteristic
Boiling Point 139° to 140°C
Melting Point −70°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapour.