Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethyl-3-heptanol, erythro + threo, 98%

$39.18 - $149.92

Chemical Identifiers

Chemical Name or Material 2,4-Dimethyl-3-heptanol
CAS 19549-72-5
Name Note erythro + threo
Molecular Formula C9H20O
Formula Weight 144.25
MDL Number MFCD00027148
Synonym 2-Pentyl isopropyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2015306

  SDS
alfa aesar™
B2015306
5g Each for $39.18

AAB2015314

  SDS
alfa aesar™
B2015314
25g Each for $149.92
Description & Specifications

Specifications

Formula Weight 144.25
Chemical Name or Material 2,4-Dimethyl-3-heptanol
Name Note erythro + threo
Assay 98%
Molecular Formula C9H20O
MDL Number MFCD00027148
CAS 19549-72-5
Quantity 5g
Synonym 2-Pentyl isopropyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.