Please login to your online account to display your discounted pricing

(2,4-dimethyl-3-furyl)methylamine, Maybridge

Chemical Identifiers

Chemical Name or Material (2, 4-dimethyl-3-furyl)methylamine
Assay 95%
Molecular Formula C7H11NO
Formula Weight 125.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC65813CB

  SDS
fisher scientific company
CC65813CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC65813DA

  SDS
fisher scientific company
CC65813DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.17
Quantity 250mg
Assay 95%
Chemical Name or Material (2, 4-dimethyl-3-furyl)methylamine
Packaging Amber Glass Bottle
Molecular Formula C7H11NO
CAS 1023814-75-6
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger