Please login to your online account to display your discounted pricing

(2,4-dimethyl-3-furyl)methylamine, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC65813DA

 View more versions of this product

Catalog No. CC65813DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 125.17
Quantity 1g
Assay 95%
Chemical Name or Material (2, 4-dimethyl-3-furyl)methylamine
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger
CAS 1023814-75-6
Molecular Formula C7H11NO

Specifications

Chemical Name or Material (2, 4-dimethyl-3-furyl)methylamine
Assay 95%
Molecular Formula C7H11NO
Formula Weight 125.17
Accessories

GHS Signal Word: Danger