Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethyl-2-pentanol, 96%

Chemical Identifiers

Chemical Name or Material 2,4-Dimethyl-2-pentanol
CAS 625-06-9
Molecular Formula C7H16O
Formula Weight 116.2
MDL Number MFCD00021811
UN Number UN1987
Beilstein 1731623
Synonym Isobutyl dimethyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0879503

 
alfa aesar™
L0879503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0879506

 
alfa aesar™
L0879506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.2
Chemical Name or Material 2,4-Dimethyl-2-pentanol
Molecular Formula C7H16O
CAS 625-06-9
Synonym Isobutyl dimethyl carbinol
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
MDL Number MFCD00021811
UN Number UN1987
Beilstein 1731623
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Quantity 1g
Flash Point 38°C (100°F)
Boiling Point 132° to 134°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.