Please login to your online account to display your discounted pricing

2,4-Dimethyl-1,3-thiazole-5-carboxylic acid, 97%


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC05801EA

 View more versions of this product

Catalog No. CC05801EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 157.19
Quantity 10g
Assay 97%
Packaging Amber Glass Bottle
Chemical Name or Material 2, 4-Dimethyl-1, 3-thiazole-5-carboxylic acid
Assay Percent Range 97%
Molecular Formula C6H7NO2S
CAS 53137-27-2

Specifications

Chemical Name or Material 2, 4-Dimethyl-1, 3-thiazole-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7NO2S
Formula Weight 157.19