missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dimethyl-1,3-dioxane 98.0+%, TCI America™
$273.73 - $459.54
Chemical Identifiers
CAS | 766-20-1 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00023821 |
InChI Key | VREPYGYMOSZTKJ-UHFFFAOYNA-N |
Synonym | 2,4-dimethyl-m-dioxane, 1,3-dioxane, 2,4-dimethyl, m-dioxane, 2,4-dimethyl, cis-2,4-dimethyl-1,3-dioxane, acmc-209p4w, 5-19-01-00072 beilstein handbook reference, 2,4-dimethyl-1,3 dioxane, 1,3-dioxane,2,4-dimethyl, trans-2,4-dimethyl-1,3-dioxane, m-dioxane, 2,4-dimethyl-8ci |
PubChem CID | 13009 |
IUPAC Name | 2,4-dimethyl-1,3-dioxane |
SMILES | CC1CCOC(C)O1 |
Chemical Identifiers
766-20-1 | |
116.16 | |
VREPYGYMOSZTKJ-UHFFFAOYNA-N | |
13009 | |
CC1CCOC(C)O1 |
C6H12O2 | |
MFCD00023821 | |
2,4-dimethyl-m-dioxane, 1,3-dioxane, 2,4-dimethyl, m-dioxane, 2,4-dimethyl, cis-2,4-dimethyl-1,3-dioxane, acmc-209p4w, 5-19-01-00072 beilstein handbook reference, 2,4-dimethyl-1,3 dioxane, 1,3-dioxane,2,4-dimethyl, trans-2,4-dimethyl-1,3-dioxane, m-dioxane, 2,4-dimethyl-8ci | |
2,4-dimethyl-1,3-dioxane |
Specifications
766-20-1 | |
118°C | |
MFCD00023821 | |
2707 | |
VREPYGYMOSZTKJ-UHFFFAOYNA-N | |
2,4-dimethyl-1,3-dioxane | |
13009 | |
≥98.0% (GC) | |
2,4-Dimethyl-1,3-dioxane |
Colorless | |
C6H12O2 | |
10 mL | |
2,4-dimethyl-m-dioxane, 1,3-dioxane, 2,4-dimethyl, m-dioxane, 2,4-dimethyl, cis-2,4-dimethyl-1,3-dioxane, acmc-209p4w, 5-19-01-00072 beilstein handbook reference, 2,4-dimethyl-1,3 dioxane, 1,3-dioxane,2,4-dimethyl, trans-2,4-dimethyl-1,3-dioxane, m-dioxane, 2,4-dimethyl-8ci | |
CC1CCOC(C)O1 | |
116.16 | |
116.16 | |
Liquid |
Safety and Handling
EINECSNumber : (5)-0838
RTECSNumber : JH0675000
TSCA : No