missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2,4-Dimethoxypyrimidine, 99+%, ACROS Organics™
Chemical Identifiers
CAS |
3551-55-1 |
Molecular Formula |
C6H8N2O2 |
Molecular Weight (g/mol) |
140.14 |
MDL Number |
MFCD00056028 |
InChI Key |
KEVRHVMWBKFGLO-UHFFFAOYSA-N |
Synonym |
2,4-dimethoxy-pyrimidine, 2,4-dimethoxypyrimidine, acmc-1adzt, dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa, ksc911k2l, pubchem7104, pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, zlchem 892 |
PubChem CID |
137976 |
IUPAC Name |
2,4-dimethoxypyrimidine |
SMILES |
COC1=NC(=NC=C1)OC |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
AC296550025
SDS
View Documents
|
Thermo Fisher Scientific
296550025
|
2.5g |
Glass bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
Description
Specifications
Chemical Identifiers
|
3551-55-1 |
|
140.14 |
|
KEVRHVMWBKFGLO-UHFFFAOYSA-N |
|
137976 |
|
COC1=NC(=NC=C1)OC |
|
C6H8N2O2 |
|
MFCD00056028 |
|
2,4-dimethoxy-pyrimidine, 2,4-dimethoxypyrimidine, acmc-1adzt, dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa, ksc911k2l, pubchem7104, pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, zlchem 892 |
|
2,4-dimethoxypyrimidine |
Specifications
|
Authentic |
|
1.5000 to 1.502 |
|
Colorless to Brown |
|
3551-55-1 |
|
100.0 |
|
C6H8N2O2 |
|
2,4-dimethoxy-pyrimidine, 2,4-dimethoxypyrimidine, acmc-1adzt, dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa, ksc911k2l, pubchem7104, pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, zlchem 892 |
|
KEVRHVMWBKFGLO-UHFFFAOYSA-N |
|
2,4-dimethoxypyrimidine |
|
137976 |
|
Liquid |
|
2, 4-Dimethoxypyrimidine |
|
Glass bottle |
|
204°C to 205°C |
|
2.5g |
|
99.0 |
|
99% min. (GC) |
|
MFCD00056028 |
|
Solubility in water: .. Other solubilities: soluble in ethanol |
|
COC1=NC(=NC=C1)OC |
|
140.14 |
|
140.14 |
|
99+% |
SDS