Please login to your online account to display your discounted pricing

2,4-Dimethoxypyrimidine, +99%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  296550025

 View more versions of this product

Catalog No. AC296550025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 140.14
Quantity 2.5g
Color Clear colorless to light brown
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2, 4-Dimethoxypyrimidine
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula C6 H8 N2 O2
CAS 3551-55-1
Infrared Spectrum Authentic
Refractive Index 1.5000 to 1.5020 (20°C, 589nm)
MDL Number MFCD00056028

Specifications

Chemical Name or Material 2, 4-Dimethoxypyrimidine
CAS 3551-55-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6 H8 N2 O2
Formula Weight 140.14
MDL Number MFCD00056028