Learn More
2,4-Dimethoxypyrimidine-5-boronic acid, 95%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC445420010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Dimethoxypyrimidine-5-boronic acid | |
White to Yellow | |
94% min. (HPLC) | |
94 % min. consistent with structure (1H-NMR) | |
2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronicacid, 5-borono-2,3-dimethoxypyrimidine | |
LKGKUACPLXCVOF-UHFFFAOYSA-N | |
2736209 | |
Crystals or Crystalline Powder |
89641-18-9 | |
107°C to 109°C | |
C6H9BN2O4 | |
Glass Bottle | |
1g | |
183.96 | |
183.96 | |
95% |
Chemical Identifiers
89641-18-9 | |
183.96 | |
2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronicacid, 5-borono-2,3-dimethoxypyrimidine |
C6H9BN2O4 | |
LKGKUACPLXCVOF-UHFFFAOYSA-N | |
2736209 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only