missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethoxypyrimidine-5-boronic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 89641-18-9
Molecular Formula C6H9BN2O4
Molecular Weight (g/mol) 183.96
InChI Key LKGKUACPLXCVOF-UHFFFAOYSA-N
Synonym 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 5-borono-2,3-dimethoxypyrimidine, 2,4-dimethoxypyrimidine-5-boronicacid
PubChem CID 2736209
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC445420010 SDS View Documents Acros Organics
AC445420010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445420050 SDS View Documents Acros Organics
AC445420050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

89641-18-9
183.96
2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 5-borono-2,3-dimethoxypyrimidine, 2,4-dimethoxypyrimidine-5-boronicacid
C6H9BN2O4
LKGKUACPLXCVOF-UHFFFAOYSA-N
2736209

Specifications

94 % min. consistent with structure (1H-NMR)
White to Yellow
89641-18-9
C6H9BN2O4
LKGKUACPLXCVOF-UHFFFAOYSA-N
2736209
Crystals or Crystalline Powder
2, 4-Dimethoxypyrimidine-5-boronic acid
Glass bottle
1g
95%
2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 5-borono-2,3-dimethoxypyrimidine, 2,4-dimethoxypyrimidine-5-boronicacid
183.96
183.96
≥94.0% (HPLC)