missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dimethoxypyrimidine-5-boronic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445420050
Specifications
2, 4-Dimethoxypyrimidine-5-boronic acid | |
Glass Bottle | |
107°C to 109°C | |
89641-18-9 | |
C6H9BN2O4 | |
LKGKUACPLXCVOF-UHFFFAOYSA-N | |
2736209 | |
Crystals or Crystalline Powder |
94 % min. consistent with structure (1H-NMR) | |
White to Yellow | |
5g | |
94% min. (HPLC) | |
2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronicacid, 5-borono-2,3-dimethoxypyrimidine | |
183.96 | |
183.96 | |
95% |
Chemical Identifiers
89641-18-9 | |
183.96 | |
2,4-dimethoxy-5-pyrimidineboronic acid, 2,4-dimethoxy-5-pyrimidinylboronic acid, 2,4-dimethoxyprimidine-5-boronic acid, 2,4-dimethoxypryimidine-5-boronic acid, 2,4-dimethoxypyrimidin-5-yl boronic acid, 2,4-dimethoxypyrimidin-5-ylboronic acid, 2,4-dimethoxypyrimidine-5-boronic acid, 2,4-dimethoxypyrimidine-5-boronicacid, 5-borono-2,3-dimethoxypyrimidine |
C6H9BN2O4 | |
LKGKUACPLXCVOF-UHFFFAOYSA-N | |
2736209 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning