Please login to your online account to display your discounted pricing

2,4-Dimethoxyphenylboronic acid, 97%, ACROS Organics™

$51.00 - $51.00

Chemical Identifiers

Chemical Name or Material 2, 4-Dimethoxybenzeneboronic acid
CAS 133730-34-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8 H11 B O4
Formula Weight 181.98
MDL Number MFCD01074590
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359200010

  SDS
thermo fisher scientific
35920-0010
Glass bottle 1g Each for $51.00
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 181.98
Physical Form Moist Powder
Quantity 1g
Color White to beige or pink
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8 H11 B O4
Assay Percent Range 97%
Chemical Name or Material 2, 4-Dimethoxybenzeneboronic acid
CAS 133730-34-4
MDL Number MFCD01074590
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.