missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxyphenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1258406

 View more versions of this product

Catalog No. AAL1258406



Specifications

Specifications

2,4-Dimethoxyphenyl isothiocyanate
166°C to 167°C (6mmHg)
5g
C9H9NO2S
UN2923
2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
COC1=CC(=C(C=C1)N=C=S)OC
195.236
195.24
>110°C (230°F)
51°C to 53°C
33904-03-9
MFCD00041058
2723226
CNXSCEPTSXPBTP-UHFFFAOYSA-N
1-isothiocyanato-2,4-dimethoxybenzene
2736202
97%

Chemical Identifiers

33904-03-9
195.236
CNXSCEPTSXPBTP-UHFFFAOYSA-N
2736202
COC1=CC(=C(C=C1)N=C=S)OC
C9H9NO2S
MFCD00041058
2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
1-isothiocyanato-2,4-dimethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents