missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethoxyphenyl isothiocyanate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441780050

 View more versions of this product

Catalog No. AC441780050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Dimethoxyphenyl isothiocyanate
Authentic
Yellow
5g
94.0
94% min. (GC)
(CH3O)2C6H3NCS
CNXSCEPTSXPBTP-UHFFFAOYSA-N
1-isothiocyanato-2,4-dimethoxybenzene
2736202
Powder
113°C
166.0°C to 167.0°C (6.0mmHg)
54.0°C to 55.0°C
33904-03-9
100.0
C9H9NO2S
1-isothiocyanato-2,4-dimethoxy-benzene, 1-isothiocyanato-2.4-dimethoxybenzene, 2,4-dimethoxybenzenisothiocyanate, 2,4-dimethoxyphenyl isothiocyanate, 2,4-dimethoxyphenylisothiocyanate, acmc-20aock, isothiocyanic acid 2,4-dimethoxyphenyl ester
COC1=CC(=C(C=C1)N=C=S)OC
195.24
195.24
95%

Chemical Identifiers

33904-03-9
195.24
1-isothiocyanato-2,4-dimethoxy-benzene, 1-isothiocyanato-2.4-dimethoxybenzene, 2,4-dimethoxybenzenisothiocyanate, 2,4-dimethoxyphenyl isothiocyanate, 2,4-dimethoxyphenylisothiocyanate, acmc-20aock, isothiocyanic acid 2,4-dimethoxyphenyl ester
1-isothiocyanato-2,4-dimethoxybenzene
C9H9NO2S
CNXSCEPTSXPBTP-UHFFFAOYSA-N
2736202
COC1=CC(=C(C=C1)N=C=S)OC
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

SDS Documents