Please login to your online account to display your discounted pricing

2,4-Dimethoxyphenyl isocyanate, 97%, ACROS Organics™


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  AC31429DA

 View more versions of this product

Catalog No. AC31429DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 179.17
Quantity 1g
Color Clear light yellow
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5465 to 1.5485 (20°C, 589nm)
Chemical Name or Material 2, 4-Dimethoxyphenyl isocyanate
Infrared Spectrum Authentic
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H9NO3
Linear Formula (CH3O)C6H3NCO
MDL Number MFCD00002005
CAS 84370-87-6
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 4-Dimethoxyphenyl isocyanate
CAS 84370-87-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9NO3
Linear Formula (CH3O)C6H3NCO
Formula Weight 179.17
MDL Number MFCD00002005
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.