missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dimethoxybenzyl alcohol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 170461000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Dimethoxybenzyl alcohol | |
98.5 | |
177.0°C to 179.0°C (10.0mmHg) | |
Authentic | |
Glass bottle | |
(CH3O)2C6H3CH2OH | |
MFCD00004614 | |
06, I, 550 | |
RNKOUSCCPHSCFE-UHFFFAOYSA-N | |
(2,4-dimethoxyphenyl)methanol | |
81750 | |
99% |
7314-44-5 | |
100.0 | |
>110°C | |
98.5% min. (GC) | |
C9H12O3 | |
1.5450 to 1.5480 | |
100g | |
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol | |
COC1=CC(=C(C=C1)CO)OC | |
168.19 | |
168.19 |
Chemical Identifiers
7314-44-5 | |
168.19 | |
RNKOUSCCPHSCFE-UHFFFAOYSA-N | |
81750 | |
COC1=CC(=C(C=C1)CO)OC |
C9H12O3 | |
MFCD00004614 | |
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol | |
(2,4-dimethoxyphenyl)methanol |
Safety and Handling
EINECSNumber : 230-775-2
RUO â Research Use Only