missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethoxybenzoic Acid 99%, ACROS Organics™

$73.45 - $73.45

Chemical Identifiers

CAS 91-52-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H10O4
Linear Formula (CH3O)2C6H3CO2H
Formula Weight 182.18
MDL Number MFCD00002434
Chemical Name or Material 2, 4-Dimethoxybenzoic acid, 99%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC115430250 SDS Thermo Fisher Scientific
115430250
Glass bottle 25g Each for $73.45
Specifications Description & Specifications

Chemical Identifiers

91-52-1
100.0
C9H10O4
182.18
2, 4-Dimethoxybenzoic acid, 99%
96.0
97%
(CH3O)2C6H3CO2H
MFCD00002434
≥97.5%

Specifications

91-52-1
100.0
Glass bottle
97%
107° to 111°C
White to off-white
MFCD00002434
2, 4-Dimethoxybenzoic acid, 99%
96.0
Authentic
Powder
C9H10O4
(CH3O)2C6H3CO2H
182.18
25g
≥97.5%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-074-1

TSCA : TSCA