Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxybenzeneboronic acid, 98%

$62.23 - $626.81

Chemical Identifiers

Chemical Name or Material 2,4-Dimethoxybenzeneboronic acid
CAS 133730-34-4
Molecular Formula C8H11BO4
Formula Weight 181.98
MDL Number MFCD01074590
Beilstein 4744213
Synonym 2,4-Dimethoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2437403

  SDS
alfa aesar™
B2437403
1g Each for $62.23

AAB2437406

  SDS
alfa aesar™
B2437406
5g Each for $152.19

AAB2437414

  SDS
alfa aesar™
B2437414
25g Each for $626.81
Description & Specifications

Specifications

Formula Weight 181.98
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Synonym 2,4-Dimethoxyphenylboronic acid
Chemical Name or Material 2,4-Dimethoxybenzeneboronic acid
Quantity 1g
Melting Point 115° to 120°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 133730-34-4
Assay 98%
Molecular Formula C8H11BO4
Beilstein 4744213
MDL Number MFCD01074590
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.