Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxybenzaldoxime, 97%

$324.78 - $324.78

Chemical Identifiers

Chemical Name or Material 2,4-Dimethoxybenzaldoxime
CAS 31874-34-7
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00082783
Beilstein 3198474
Synonym 2,4-Dimethoxybenzaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0989414

 
alfa aesar™
L0989414
25g Each for $324.78
Description & Specifications

Specifications

Formula Weight 181.19
Molecular Formula C9H11NO3
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2,4-Dimethoxybenzaldoxime
MDL Number MFCD00082783
CAS 31874-34-7
Melting Point 105° to 108°C
Beilstein 3198474
Quantity 25g
Synonym 2,4-Dimethoxybenzaldehyde oxime
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.