missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxybenzaldoxime, 97%

Manufacturer:  Alfa Aesar™ L0989406

 View more versions of this product

Catalog No. AAL0989406



Specifications

Specifications

2,4-Dimethoxybenzaldoxime
5g
C9H11NO3
3198474
SFDRVCQSVTYHLU-UXBLZVDNSA-N
(NE)-N-[(2,4-dimethoxyphenyl)methylidene]hydroxylamine
6871293
97%
105°C to 108°C
31874-34-7
MFCD00082783
2,4-dimethoxybenzaldoxime, 2,4-dimethoxybenzaldehyde oxime, e-n-2,4-dimethoxyphenyl methylidene hydroxylamine, benzaldehyde, 2,4-dimethoxy-, oxime, 2.4-dimethoxybenzaldoxim, e-2,4-dimethoxybenzaldehyde oxime, 2,4-dimethoxyphenyl hydroxyimino methane, ne-n-2,4-dimethoxyphenyl methylidene hydroxylamine
COC1=CC(=C(C=C1)C=NO)OC
181.191
181.19

Chemical Identifiers

31874-34-7
181.191
SFDRVCQSVTYHLU-UXBLZVDNSA-N
6871293
COC1=CC(=C(C=C1)C=NO)OC
C9H11NO3
MFCD00082783
2,4-dimethoxybenzaldoxime, 2,4-dimethoxybenzaldehyde oxime, e-n-2,4-dimethoxyphenyl methylidene hydroxylamine, benzaldehyde, 2,4-dimethoxy-, oxime, 2.4-dimethoxybenzaldoxim, e-2,4-dimethoxybenzaldehyde oxime, 2,4-dimethoxyphenyl hydroxyimino methane, ne-n-2,4-dimethoxyphenyl methylidene hydroxylamine
(NE)-N-[(2,4-dimethoxyphenyl)methylidene]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents