missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethoxyaniline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115350050

 View more versions of this product

Catalog No. AC115350050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 4-Dimethoxyaniline
Authentic
Brown
5g
2735-04-8
100.0
C8H11NO2
MFCD00008371
2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline, 2,4-dimethoxy-anilin, 2,4-dimethoxyphenylamine, 2,4-domethoxyaniline, aniline, 2,4-dimethoxy, benzenamine, 2,4-dimethoxy, benzenamine, 2,4-dimethoxy-, hydrochloride, unii-i079354gsi
GEQNZVKIDIPGCO-UHFFFAOYSA-N
2,4-dimethoxyaniline
17652
Crystalline Solid
>110°C
Glass bottle
34°C to 37°C
97%
96.0
96% min. (GC)
(CH3O)2C6H3NH2
13, 784
Solubility in water: 7g/L (20°C)
COC1=CC(=C(C=C1)N)OC
153.18
153.18
97%

Chemical Identifiers

2735-04-8
153.18
GEQNZVKIDIPGCO-UHFFFAOYSA-N
17652
COC1=CC(=C(C=C1)N)OC
C8H11NO2
MFCD00008371
2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline, 2,4-dimethoxy-anilin, 2,4-dimethoxyphenylamine, 2,4-domethoxyaniline, aniline, 2,4-dimethoxy, benzenamine, 2,4-dimethoxy, benzenamine, 2,4-dimethoxy-, hydrochloride, unii-i079354gsi
2,4-dimethoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-355-7

RTECSNumber : BX4200000

TSCA : TSCA

SDS Documents