Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxyaniline, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dimethoxyaniline
CAS 2735-04-8
Molecular Formula C8H11NO2
Formula Weight 153.18
MDL Number MFCD00008371
UN Number UN2811
Beilstein 638704
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1394318

  SDS
alfa aesar™
A1394318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1394330

  SDS
alfa aesar™
A1394330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.18
Chemical Name or Material 2,4-Dimethoxyaniline
TSCA TSCA
Assay 97%
Molecular Formula C8H11NO2
Melting Point 32° to 37°C
MDL Number MFCD00008371
CAS 2735-04-8
UN Number UN2811
Beilstein 638704
Quantity 50g
Flash Point >110°C (230°F)
Odor Characteristic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H331-H373-H302
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number BX4200000

Applications
Various 2,4-dimethoxyphenylsemicarbazones were synthesized starting from 2,4-dimethoxyaniline via a phenylcarbamate intermediate. The hydrochloride of 2,4-dimethoxyaniline was prepared by intro- ducing hydrogen chloride gas into an etheral solution and washing the precipitate with absolute diethyl ether.

Solubility
Slightly soluble in water(7g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. May be incompatible with isocyanates, halogenated organics, peroxides, phenols (acidic), epoxides, anhydrides, and acid halides.

Safety and Handling

Danger

GHS P Statement
P280h-P309-P310-P302+P352
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H331-H373-H302
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.