Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxy-beta-nitrostyrene, 99%

$116.07 - $116.07

Chemical Identifiers

Chemical Name or Material 2,4-Dimethoxy-beta-nitrostyrene
CAS 1891-10-7
Molecular Formula C10H11NO4
Formula Weight 209.19
MDL Number MFCD00015922
Synonym 1-(2,4-Dimethoxyphenyl)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2056406

  SDS
alfa aesar™
B2056406
5g Each for $116.07
Description & Specifications

Specifications

Formula Weight 209.19
Chemical Name or Material 2,4-Dimethoxy-beta-nitrostyrene
Molecular Formula C10H11NO4
Melting Point 99° to 101°C
CAS 1891-10-7
Quantity 5g
Synonym 1-(2,4-Dimethoxyphenyl)-2-nitroethylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
MDL Number MFCD00015922
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.