Please login to your online account to display your discounted pricing

2,4-Dimethoxy-4'-fluorochalcone, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dimethoxy-4'-fluorochalcone
Name Note 97%
CAS 336101-27-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C17H15FO3
Linear Formula (CH3O)2C6H3CHCHC(O)C6H4F
Formula Weight 286.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC339970250

 
thermo fisher scientific
339970250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 286.3
Physical Form Powder
Quantity 25g
Color Light yellow
TSCA TSCA
Melting Point 117° to 122°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C17H15FO3
CAS 336101-27-0
Chemical Name or Material 2, 4-Dimethoxy-4'-fluorochalcone
Linear Formula (CH3O)2C6H3CHCHC(O)C6H4F
Assay Percent Range 97%
Name Note 97%
MDL Number MFCD01901412
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.