Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethoxy-1-nitrobenzene, 97%

$58.15 - $221.50

Chemical Identifiers

Chemical Name or Material 2,4-Dimethoxy-1-nitrobenzene
CAS 4920-84-7
Molecular Formula C8H9NO4
Formula Weight 183.16
MDL Number MFCD00024210
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2535306

  SDS
alfa aesar™
B2535306
5g Each for $58.15

AAB2535314

  SDS
alfa aesar™
B2535314
25g Each for $221.50
Description & Specifications

Specifications

Formula Weight 183.16
Chemical Name or Material 2,4-Dimethoxy-1-nitrobenzene
Assay 97%
Molecular Formula C8H9NO4
MDL Number MFCD00024210
CAS 4920-84-7
Melting Point 75° to 77°C
Quantity 5g