missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diisopropylphenol, 98%

Manufacturer:  Alfa Aesar™ H5502606

 View more versions of this product

Catalog No. AAH5502606



Description

Description

Applications
2,4-Diisopropylphenol is used as an antioxidant.

Solubility
Sparingly soluble at water (0.25 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2,4-Diisopropylphenol
79°C to 80°C
2942-06-5
MFCD01707536
QLUFBCVWKTWKBF-UHFFFAOYSA-N
6-nitro-1,3-benzothiazole
76256
98%
110°C (230°F)
5g
C7H4N2O2S
#
C1=CC2=C(C=C1[N+](=O)[O-])SC=N2
180.18
178.28

Chemical Identifiers

2942-06-5
180.18
QLUFBCVWKTWKBF-UHFFFAOYSA-N
76256
C1=CC2=C(C=C1[N+](=O)[O-])SC=N2
C7H4N2O2S
MFCD01707536
#
6-nitro-1,3-benzothiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SL0800000

Documents