missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dihydroxypyrimidine-5-carboxylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 155200050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Dihydroxypyrimidine-5-carboxylic acid | |
23945-44-0 | |
100.0 | |
97.5% min. (HPLC) | |
C5H4N2O4 | |
5g | |
2,4-dihydroxypyrimidine-5-carboxylic acid, uracil 5-carboxylic acid, isoorotic acid, uracil-5-carboxylic acid, 5-carboxyuracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid, 2,4-dihydroxy-5-pyrimidinecarboxylic acid, 5-uracilcarboxylic acid, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo, uracil 5-carboxylate | |
C1=C(C(=O)NC(=O)N1)C(=O)O | |
156.1 | |
CHEBI:17477 | |
98% |
98% | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00149397 | |
25, 256 | |
ZXYAAVBXHKCJJB-UHFFFAOYSA-N | |
2,4-dioxo-1H-pyrimidine-5-carboxylic acid | |
90301 | |
156.1 |
Safety and Handling
EINECSNumber : 245-947-2
TSCA : TSCA
RUO â Research Use Only