missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dihydroxypyrimidine-5-carboxylic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 155200250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 4-Dihydroxypyrimidine-5-carboxylic acid | |
23945-44-0 | |
100.0 | |
White to Yellow | |
97.5% min. (HPLC) | |
C5H4N2O4 | |
25 g | |
2,4-dihydroxypyrimidine-5-carboxylic acid, uracil 5-carboxylic acid, isoorotic acid, uracil-5-carboxylic acid, 5-carboxyuracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid, 2,4-dihydroxy-5-pyrimidinecarboxylic acid, 5-uracilcarboxylic acid, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo, uracil 5-carboxylate | |
C1=C(C(=O)NC(=O)N1)C(=O)O | |
156.1 | |
CHEBI:17477 | |
98% |
98% | |
97.5 | |
283.0°C | |
Authentic | |
Glass bottle | |
MFCD00149397 | |
25, 256 | |
ZXYAAVBXHKCJJB-UHFFFAOYSA-N | |
2,4-dioxo-1H-pyrimidine-5-carboxylic acid | |
90301 | |
156.1 | |
Crystalline Powder |
Chemical Identifiers
23945-44-0 | |
156.1 | |
ZXYAAVBXHKCJJB-UHFFFAOYSA-N | |
90301 | |
2,4-dioxo-1H-pyrimidine-5-carboxylic acid |
C5H4N2O4 | |
MFCD00149397 | |
2,4-dihydroxypyrimidine-5-carboxylic acid, uracil 5-carboxylic acid, isoorotic acid, uracil-5-carboxylic acid, 5-carboxyuracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid, 2,4-dihydroxy-5-pyrimidinecarboxylic acid, 5-uracilcarboxylic acid, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo, uracil 5-carboxylate | |
CHEBI:17477 | |
C1=C(C(=O)NC(=O)N1)C(=O)O |
Safety and Handling
EINECSNumber : 245-947-2
TSCA : TSCA
RUO – Research Use Only