Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dihydroxypteridine, 98%

$24.64 - $112.51

Chemical Identifiers

Chemical Name or Material 2,4-Dihydroxypteridine
CAS 487-21-8
Molecular Formula C6H4N4O2
Formula Weight 164.12
MDL Number MFCD00066250
Beilstein 157503
Merck Index 14,5596
Synonym Lumazine; 2,4-Pteridinediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0673203

 
alfa aesar™
L0673203
1g Each for $24.64

AAL0673206

 
alfa aesar™
L0673206
5g Each for $112.51
Description & Specifications

Specifications

Formula Weight 164.12
Chemical Name or Material 2,4-Dihydroxypteridine
CAS 487-21-8
TSCA TSCA
Assay 98%
Molecular Formula C6H4N4O2
MDL Number MFCD00066250
Merck Index 14,5596
Synonym Lumazine; 2,4-Pteridinediol
Melting Point ≈348°C
Quantity 1g
Beilstein 157503
RTECS Number UO3416000