Please login to your online account to display your discounted pricing

2,4-Dihydroxybenzophenone 99%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  155185000

 View more versions of this product

Catalog No. AC155185000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 214.22
Quantity 500g
Color Yellow
Melting Point 144° to 147°C
TSCA TSCA
RTECS Number DJ0700000
EINECS Number 205-029-4
Merck Index 15, 1109
Beilstein 08, 312
CAS Max % 100.0
Synonym Benzoresorcinol
CAS Min % 98.5
Packaging Plastic bottle
Infrared Spectrum Authentic
Loss on Drying 0.1% max. ()
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Linear Formula (HO)2C6H3COC6H5
CAS 131-56-6
Chemical Name or Material 2, 4-Dihydroxybenzophenone
Molecular Formula C13H10O3
Name Note 99%
MDL Number MFCD00002277

Specifications

Chemical Name or Material 2, 4-Dihydroxybenzophenone
Name Note 99%
CAS 131-56-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H10O3
Linear Formula (HO)2C6H3COC6H5
Formula Weight 214.22
MDL Number MFCD00002277
Synonym Benzoresorcinol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents