Please login to your online account to display your discounted pricing

2,4-Dihydroxybenzophenone 99%, ACROS Organics™

$133.66 - $133.66

Chemical Identifiers

Chemical Name or Material 2, 4-Dihydroxybenzophenone
Name Note 99%
CAS 131-56-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H10O3
Linear Formula (HO)2C6H3COC6H5
Formula Weight 214.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155181000

  SDS
fisher scientific acros
155181000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155185000

  SDS
thermo fisher scientific
155185000
Plastic bottle 500g Each for $133.66
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 214.22
Quantity 100g
Color Yellow
Beilstein 08, 312
Merck Index 15, 1109
TSCA TSCA
EINECS Number 205-029-4
RTECS Number DJ0700000
Melting Point 144° to 147°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS Min % 98.5
Synonym Benzoresorcinol
Loss on Drying 0.1% max. ()
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material 2, 4-Dihydroxybenzophenone
Molecular Formula C13H10O3
Linear Formula (HO)2C6H3COC6H5
MDL Number MFCD00002277
Assay Percent Range 99%
CAS 131-56-6
Name Note 99%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.