Learn More
2,4-Dihydroxybenzoic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114871000
Specifications
2, 4-Dihydroxybenzoic acid | |
Plastic bottle | |
210.0°C to 215.0°C | |
97% | |
96.0 | |
97% | |
(HO)2C6H3CO2H | |
10, 377 | |
2,4-dhba, 2,4-dihydroxy-benzoic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, b-resorcylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, beta-resorcylic acid, p-hydroxysalicylic acid, resorcylic acid, beta | |
UIAFKZKHHVMJGS-UHFFFAOYSA-N | |
2,4-dihydroxybenzoic acid | |
1491 | |
Crystalline Powder |
Authentic | |
Beige to Cream or Pink | |
100g | |
89-86-1 | |
100.0 | |
C7H6O4 | |
MFCD00002451 | |
15, 8278 | |
Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol and ether,practically insoluble in benzene | |
C1=CC(=C(C=C1O)O)C(=O)O | |
154.12 | |
154.12 | |
97% |
Chemical Identifiers
89-86-1 | |
154.12 | |
UIAFKZKHHVMJGS-UHFFFAOYSA-N | |
1491 | |
C1=CC(=C(C=C1O)O)C(=O)O |
C7H6O4 | |
MFCD00002451 | |
2,4-dhba, 2,4-dihydroxy-benzoic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, b-resorcylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, beta-resorcylic acid, p-hydroxysalicylic acid, resorcylic acid, beta | |
2,4-dihydroxybenzoic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 201-946-9
RTECSNumber : VH3708050
TSCA : TSCA