Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dihydroxybenzhydrazide, 95%

$438.47 - $438.47

Chemical Identifiers

Chemical Name or Material 2,4-Dihydroxybenzhydrazide
CAS 13221-86-8
Molecular Formula C7H8N2O3
Formula Weight 168.15
MDL Number MFCD00017053
Beilstein 2641320
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0991814

  SDS
alfa aesar™
L0991814
25g Each for $438.47
Description & Specifications

Specifications

Formula Weight 168.15
Chemical Name or Material 2,4-Dihydroxybenzhydrazide
CAS 13221-86-8
MDL Number MFCD00017053
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H8N2O3
Quantity 25g
Assay 95%
Melting Point 240° to 242°C
Health Hazard 1 Warning
Beilstein 2641320
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.