missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dihydroxybenzhydrazide, 95%

Manufacturer:  Alfa Aesar™ L0991814

 View more versions of this product

Catalog No. AAL0991814



Specifications

Specifications

2,4-Dihydroxybenzhydrazide
25g
C7H8N2O3
2641320
GOPUHTXVZGIOHN-UHFFFAOYSA-N
2,4-dihydroxybenzohydrazide
166785
95%
240°C to 242°C
13221-86-8
MFCD00017053
2,4-dihydroxybenzhydrazide, 2,4-dihydroxybenzoic acid hydrazide, benzoic acid, 2,4-dihydroxy-, hydrazide, 2,4-dihydroxybenzoic hydrazide, 2,4-bis oxidanyl benzohydrazide, 2,4-dihydroxybenzoic acid, hydrazide, acmc-1cf1y, 4-???benzene-1,3-diol, 2,4-dihydroxybenzoylhydrazine, 2,4-dihydroxy benzoic acid hydrazide
C1=CC(=C(C=C1O)O)C(=O)NN
168.152
168.15

Chemical Identifiers

13221-86-8
168.152
GOPUHTXVZGIOHN-UHFFFAOYSA-N
166785
C1=CC(=C(C=C1O)O)C(=O)NN
C7H8N2O3
MFCD00017053
2,4-dihydroxybenzhydrazide, 2,4-dihydroxybenzoic acid hydrazide, benzoic acid, 2,4-dihydroxy-, hydrazide, 2,4-dihydroxybenzoic hydrazide, 2,4-bis oxidanyl benzohydrazide, 2,4-dihydroxybenzoic acid, hydrazide, acmc-1cf1y, 4-???benzene-1,3-diol, 2,4-dihydroxybenzoylhydrazine, 2,4-dihydroxy benzoic acid hydrazide
2,4-dihydroxybenzohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents