missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dihydroxybenzaldehyde, 98%

$90.81 - $1,022.08

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Formula Weight 138.12
MDL Number MFCD00011686
Beilstein 878548
Merck Index 14,8156
Synonym beta-Resorcylaldehyde
Chemical Name or Material 2,4-Dihydroxybenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1286814 SDS Alfa Aesar™
A1286814
25g Each for $90.81
AAA1286836 SDS Alfa Aesar™
A1286836
500g Each for $1,022.08
AAA1286822 SDS Alfa Aesar™
A1286822
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Description & Specifications

Chemical Identifiers

95-01-2
138.12
878548
beta-Resorcylaldehyde
C7H6O3
MFCD00011686
14,8156
2,4-Dihydroxybenzaldehyde

Specifications

220°C (22mmHg)
25g
C7H6O3
MFCD00011686
14,8156
2,4-Dihydroxybenzaldehyde
134° to 138°C
95-01-2
138.12
878548
beta-Resorcylaldehyde
98%

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VH3600000

TSCA : TSCA