Please login to your online account to display your discounted pricing

2,4-Dihydroxy-6-methylpyrimidine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dihydroxy-6-methylpyrimidine
Name Note 97%
CAS 626-48-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Formula Weight 126.11
MDL Number MFCD00006028
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114980050

  SDS
thermo fisher scientific
114980050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystalline Powder
Assay ≥96.0%
Formula Weight 126.11
Quantity 5g
Color White to almost white
EINECS Number 210-949-4
TSCA TSCA
RTECS Number YR0700000
Beilstein 24, 342
Merck Index 15, 6209
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 4-Dihydroxy-6-methylpyrimidine
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
MDL Number MFCD00006028
CAS 626-48-2
Synonym 6-Methyluracil
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic