Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dihydroxy-3-nitropyridine, 99%

$48.34 - $575.69

Chemical Identifiers

Chemical Name or Material 2,4-Dihydroxy-3-nitropyridine
CAS 89282-12-2
Molecular Formula C5H4N2O4
Formula Weight 156.1
MDL Number MFCD01075671
Beilstein 149558
Synonym 4-Hydroxy-3-nitro-2-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1493303

  SDS
alfa aesar™
L1493303
1g Each for $48.34

AAL1493306

  SDS
alfa aesar™
L1493306
5g Each for $147.92

AAL1493314

  SDS
alfa aesar™
L1493314
25g Each for $575.69
Description & Specifications

Specifications

Formula Weight 156.1
Chemical Name or Material 2,4-Dihydroxy-3-nitropyridine
CAS 89282-12-2
Beilstein 149558
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Molecular Formula C5H4N2O4
MDL Number MFCD01075671
Melting Point ≈265°C
Quantity 1g
Synonym 4-Hydroxy-3-nitro-2-pyridone
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.