missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4-Dihydroxy-3-methoxychalcone, 97%

$107.57 - $321.62

Chemical Identifiers

CAS 220430-82-0
Molecular Formula C16H14O4
Formula Weight 270.29
MDL Number MFCD03788903
Synonym 2'-Hydroxy-2-(4-hydroxy-3-methoxybenzylidene)acetophenone
Chemical Name or Material 2',4-Dihydroxy-3-methoxychalcone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2753706 SDS Alfa Aesar™
H2753706
5g Each for $321.62
AAH2753703 SDS Alfa Aesar™
H2753703
1g Each for $107.57
Specifications Description & Specifications

Chemical Identifiers

220430-82-0
270.29
2'-Hydroxy-2-(4-hydroxy-3-methoxybenzylidene)acetophenone
C16H14O4
MFCD03788903
2',4-Dihydroxy-3-methoxychalcone

Specifications

127° to 129°C
220430-82-0
270.29
2'-Hydroxy-2-(4-hydroxy-3-methoxybenzylidene)acetophenone
97%
5g
C16H14O4
MFCD03788903
2',4-Dihydroxy-3-methoxychalcone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning