Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dihydroxy-2-methoxychalcone, 97%

$102.01 - $307.38

Chemical Identifiers

Chemical Name or Material 2',4'-Dihydroxy-2-methoxychalcone
CAS 104236-78-4
Molecular Formula C16H14O4
Formula Weight 270.29
MDL Number MFCD00076017
Synonym 2',4'-Dihydroxy-2-(2-methoxybenzylidene)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2753303

  SDS
alfa aesar™
H2753303
1g Each for $102.01

AAH2753306

  SDS
alfa aesar™
H2753306
5g Each for $307.38
Description & Specifications

Specifications

Formula Weight 270.29
MDL Number MFCD00076017
Melting Point 175° to 178°C
Synonym 2',4'-Dihydroxy-2-(2-methoxybenzylidene)acetophenone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2',4'-Dihydroxy-2-methoxychalcone
Quantity 1g
Molecular Formula C16H14O4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 104236-78-4
Health Hazard 1 Warning
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.