missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone, 97%

$85.14 - $252.91

Chemical Identifiers

CAS 17720-60-4
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00498162
InChI Key KLFCJXAPIFIIFR-UHFFFAOYSA-N
Synonym 1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl ethanone, 2,4,4'-trihydroxydeoxybenzoin, 2,4,4'-trihydroxy deoxybenzoin, 1-2,4-dihydroxy-phenyl-2-4-hydroxy-phenyl-ethanone, 1-2,4-dihydroxy-phenyl 2 4-hydro xy-phenyl-ethanone, ethanone,1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl, ethanone, 1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl, 2,4,4'-terthydroxydeoxybenzoin, 2,4,4-trihydroxydeoxybenzoin, 2,4,4/'-trihydroxydeoxybenzoin
PubChem CID 640096
IUPAC Name 1-(2,4-dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone
SMILES C1=CC(=CC=C1CC(=O)C2=C(C=C(C=C2)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66063MD View Documents Alfa Aesar™
H66063MD
250mg Each for $85.14
 
AAH6606303 View Documents Alfa Aesar™
H6606303
1g Each for $252.91
 
Specifications

Chemical Identifiers

17720-60-4
244.246
KLFCJXAPIFIIFR-UHFFFAOYSA-N
640096
C1=CC(=CC=C1CC(=O)C2=C(C=C(C=C2)O)O)O
C14H12O4
MFCD00498162
1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl ethanone, 2,4,4'-trihydroxydeoxybenzoin, 2,4,4'-trihydroxy deoxybenzoin, 1-2,4-dihydroxy-phenyl-2-4-hydroxy-phenyl-ethanone, 1-2,4-dihydroxy-phenyl 2 4-hydro xy-phenyl-ethanone, ethanone,1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl, ethanone, 1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl, 2,4,4'-terthydroxydeoxybenzoin, 2,4,4-trihydroxydeoxybenzoin, 2,4,4/'-trihydroxydeoxybenzoin
1-(2,4-dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone

Specifications

250mg
C14H12O4
1288279
KLFCJXAPIFIIFR-UHFFFAOYSA-N
1-(2,4-dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone
640096
Solid
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone
17720-60-4
MFCD00498162
1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl ethanone, 2,4,4'-trihydroxydeoxybenzoin, 2,4,4'-trihydroxy deoxybenzoin, 1-2,4-dihydroxy-phenyl-2-4-hydroxy-phenyl-ethanone, 1-2,4-dihydroxy-phenyl 2 4-hydro xy-phenyl-ethanone, ethanone,1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl, ethanone, 1-2,4-dihydroxyphenyl-2-4-hydroxyphenyl, 2,4,4'-terthydroxydeoxybenzoin, 2,4,4-trihydroxydeoxybenzoin, 2,4,4/'-trihydroxydeoxybenzoin
C1=CC(=CC=C1CC(=O)C2=C(C=C(C=C2)O)O)O
244.246
244.25
97%