Please login to your online account to display your discounted pricing

2,4-Difluorotoluene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Difluorotoluene
Name Note 98%
CAS 452-76-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6F2
Linear Formula CH3C6H3F2
Formula Weight 128.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC232210250

  SDS
thermo fisher scientific
232210250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC232210050

  SDS
thermo fisher scientific
232210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 128.12
Quantity 25g
Color Clear colorless
EINECS Number 207-211-9
TSCA TSCA
Density 1.1500g/mL
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4475 to 1.4495 (20°C, 589nm)
Chemical Name or Material 2, 4-Difluorotoluene
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 452-76-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Name Note 98%
Molecular Formula C7H6F2
Linear Formula CH3C6H3F2
MDL Number MFCD00000332
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.