missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Difluorotoluene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 232210050

 View more versions of this product

Catalog No. AC232210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 4-Difluorotoluene
13°C
Glass bottle
1.15
Colorless
98%
97.5
97.5% min. (GC)
CH3C6H3F2
2, 4-difluorotoluene, 2,4-difluor-1-methylbenzol, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluorotoluene, 2,4-difluorotolune, benzene, 2,4-difluoro-1-methyl, difluorotoluene, pubchem1612, pubchem1613
CC1=C(C=C(C=C1)F)F
128.12
128.12
98%
1.1500g/mL
Authentic
1.4475 to 1.4495
114.0°C to 116.0°C
5g
452-76-6
100.0
C7H6F2
MFCD00000332
MPXDAIBTYWGBSL-UHFFFAOYSA-N
2,4-difluoro-1-methylbenzene
67983
Liquid

Chemical Identifiers

452-76-6
128.12
MPXDAIBTYWGBSL-UHFFFAOYSA-N
67983
CC1=C(C=C(C=C1)F)F
C7H6F2
MFCD00000332
2, 4-difluorotoluene, 2,4-difluor-1-methylbenzol, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluorotoluene, 2,4-difluorotolune, benzene, 2,4-difluoro-1-methyl, difluorotoluene, pubchem1612, pubchem1613
2,4-difluoro-1-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-211-9

TSCA : TSCA

SDS Documents