Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Difluorothiophenol, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Difluorothiophenol
CAS 1996-44-7
Molecular Formula C6H4F2S
Formula Weight 146.16
MDL Number MFCD00203477
UN Number UN1228
Beilstein 2516728
Synonym 2,4-Difluorobenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2472603

 
alfa aesar™
B2472603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2472606

 
alfa aesar™
B2472606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2472614

 
alfa aesar™
B2472614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.16
Assay 97%
CAS 1996-44-7
Synonym 2,4-Difluorobenzenethiol
Flash Point 53°C (127°F)
Quantity 1g
Chemical Name or Material 2,4-Difluorothiophenol
Molecular Formula C6H4F2S
MDL Number MFCD00203477
UN Number UN1228
Beilstein 2516728
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.
Boiling Point 52°C (15mmHg)
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.